Methyl 3-chloroimidazo[1,2-a]pyridine-7-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-7-(methoxycarbonyl)imidazo[1,2-a]pyridine

OR70025-1G OR70025-250MG
APOSOR70025-1GEA 186 EUR
APOSOR70025-1G APOSOR70025-250MG
Methyl 3-chloroimidazo[1,2-a]pyridine-7-carboxylate
Methyl 3-chloroimidazo[1,2-a]pyridine-7-carboxylate
Formel: C₉H₇ClN₂O₂
Molecular Weight: 210.617 g/mol
Schmelzpunkt: 147…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD21602380
CAS-Nummer: 1396554-42-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR