Ethyl [(7-chloroquinolin-4-yl)thio]acetate

Lieferant: Apollo Scientific

Synonyme: Ethyl 2-[(7-chloroquinolin-4-yl)sulphanyl]ethanoate, Ethyl [(7-chloroquinolin-4-yl)sulphanyl]acetate

OR70026-1G OR70026-10G OR70026-5G
APOSOR70026-1GEA 87 EUR
APOSOR70026-1G APOSOR70026-10G APOSOR70026-5G
Ethyl [(7-chloroquinolin-4-yl)thio]acetate
Ethyl [(7-chloroquinolin-4-yl)thio]acetate
Formel: C₁₃H₁₂ClNO₂S
Molecular Weight: 281.758 g/mol
Schmelzpunkt: 63…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD21602382
CAS-Nummer: 1071597-98-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR