3-Amino-4-methoxybenzeneboronic acid 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-boronoanisole, 5-Borono-2-methoxyaniline
OR70046-1G
OR70046-250MG
APOSOR70046-1GEA
804
EUR
APOSOR70046-1G
APOSOR70046-250MG
3-Amino-4-methoxybenzeneboronic acid 96%
3-Amino-4-methoxybenzeneboronic acid
Formel:
C₇H₁₀BNO₃ Molecular Weight: 166,97 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03095251 CAS-Nummer: 879893-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...