3-Bromo-5-methoxybenzyl bromide
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Bromo-5-(bromomethyl)anisole, 1-Bromo-3-(bromomethyl)-5-methoxybenzene, 3-Bromo-5-(bromomethyl)phenyl methyl ether
OR70061-1G
OR70061-5G
APOSOR70061-1GEA
58
EUR
APOSOR70061-1G
APOSOR70061-5G
3-Bromo-5-methoxybenzyl bromide
3-Bromo-5-methoxybenzyl bromide
Formel:
C₈H₈Br₂O Molecular Weight: 279,957 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD12761608 CAS-Nummer: 59297-29-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...