2-Bromo-3-iodophenol

Lieferant: Apollo Scientific
Achtung

OR70063-1G OR70063-5G OR70063-25G
APOSOR70063-1GEA 98.6 EUR
APOSOR70063-1G APOSOR70063-5G APOSOR70063-25G
2-Bromo-3-iodophenol
2-Bromo-3-iodophenol
Formel: C₆H₄BrIO
Molecular Weight: 298,904 g/mol
Schmelzpunkt: 69…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08166322
CAS-Nummer: 863870-88-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR