5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5,6,7,8-Tetrahydro-1,2,4-triazolo[4,3-a]pyrazine
OR70087-1G
OR70087-5G
APOSOR70087-1GEA
423
EUR
APOSOR70087-1G
APOSOR70087-5G
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine 95+%
5,6,7,8-Tetrahydro[1,2,4]triazolo[4,3-a]pyrazine
Formel:
C₅H₈N₄ Molecular Weight: 124,144 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115284 CAS-Nummer: 345311-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...