2,3'-Dimethylbiphenyl
Lieferant: Apollo Scientific
|
Achtung
|
OR70105-1G
OR70105-250MG
APOSOR70105-1GEA
255
EUR
APOSOR70105-1G
APOSOR70105-250MG
2,3'-Dimethylbiphenyl
2,3'-Dimethylbiphenyl
Formel:
C₁₄H₁₄ Molecular Weight: 182,261 g/mol Siedepunkt: 273…274 °C Dichte: 0,9984 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD22628915 CAS-Nummer: 611-43-8 |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...