2-Bromo-3-iodoanisole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Bromo-1-iodo-3-methoxybenzene, 2-Bromo-3-iodophenyl methyl ether

OR70126-1G OR70126-5G OR70126-25G
APOSOR70126-1GEA 137 EUR
APOSOR70126-1G APOSOR70126-5G APOSOR70126-25G
2-Bromo-3-iodoanisole
2-Bromo-3-iodoanisole
Formel: C₇H₆BrIO
Molecular Weight: 312,93 g/mol
Schmelzpunkt: 43…47 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD18391616
CAS-Nummer: 74128-84-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR