2-Methyl-4-sulphamoylbenzeneboronic acid, pinacol ester 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Aminosulphonyl)-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)toluene, 3-Methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzenesulphonamide
OR70129-100MG
APOSOR70129-100MGEA
128
EUR
APOSOR70129-100MG
2-Methyl-4-sulphamoylbenzeneboronic acid, pinacol ester 95%
2-Methyl-4-sulphamoylbenzeneboronic acid, pinacol ester
Formel:
C₁₃H₂₀BNO₄S Molecular Weight: 297,178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18732812 CAS-Nummer: 1261295-07-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...