(1S,2S)-(+)-2-Aminocyclohexan-1-ol

Lieferant: Apollo Scientific

Synonyme: (1S,2S)-(+)-2-Hydroxycyclohexan-1-amine, (1S,2S)-2-Aminocyclohexanol, (1S,2S)-(+)-1-Amino-2-hydroxycyclohexane

OR70141-100G OR70141-25G OR70141-5G
APOSOR70141-100GEA 836 EUR
APOSOR70141-100G APOSOR70141-25G APOSOR70141-5G
(1S,2S)-(+)-2-Aminocyclohexan-1-ol
(1S,2S)-(+)-2-Aminocyclohexan-1-ol
Formel: C₆H₁₃NO
Molecular Weight: 115,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061326
CAS-Nummer: 74111-21-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR