Methyl 3-cyano-5-nitrobenzoate

Lieferant: Apollo Scientific
Achtung

OR70185-5G
APOSOR70185-5GEA 97.3 EUR
APOSOR70185-5G
Methyl 3-cyano-5-nitrobenzoate
Methyl 3-cyano-5-nitrobenzoate
Formel: C₉H₆N₂O₄
Molecular Weight: 206,155 g/mol
Schmelzpunkt: 139…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00458621
CAS-Nummer: 99066-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR