4-(1H-Pyrazol-1-ylsulphonyl)benzeneboronic acid 98%

Lieferant: Apollo Scientific

Synonyme: (4-((1H-Pyrazol-1-yl)sulphonyl)phenyl)boronic acid

OR7026-1G OR7026-250MG
APOSOR7026-1GEA 315 EUR
APOSOR7026-1G APOSOR7026-250MG
4-(1H-Pyrazol-1-ylsulphonyl)benzeneboronic acid 98%
4-(1H-Pyrazol-1-ylsulphonyl)benzeneboronic acid
Formel: C₉H₉BN₂O₄S
Molecular Weight: 252.055 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027235
CAS-Nummer: 957061-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR