4-(2-Aminoethyl)-3,5-dimethyl-1H-pyrazole dihydrobromide
Lieferant: Apollo Scientific
Achtung
Synonyme: 2-(3,5-Dimethyl-1H-pyrazol-4-yl)ethylamine dihydrobromide
OR7060-1G OR7060-250MG
APOSOR7060-1GEA 348 EUR
APOSOR7060-1G APOSOR7060-250MG
4-(2-Aminoethyl)-3,5-dimethyl-1H-pyrazole dihydrobromide
4-(2-Aminoethyl)-3,5-dimethyl-1H-pyrazole dihydrobromide
Formel: C₇H₁₃N₃·2BrH
Molecular Weight: 301,022 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445535
CAS-Nummer: 1257658-83-5