Ethyl 4-bromo-2-methylsulphanylbenzoate 97%

Lieferant: Apollo Scientific

Synonyme: Ethyl 4-bromo-2-(methylthio)benzoate

OR71003-1G OR71003-500MG
APOSOR71003-1GEA 780 EUR
APOSOR71003-1G APOSOR71003-500MG
Ethyl 4-bromo-2-methylsulphanylbenzoate 97%
Ethyl 4-bromo-2-methylsulphanylbenzoate
Formel: C₁₀H₁₁BrO₂S
Molecular Weight: 275.162 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22488928
CAS-Nummer: 918328-04-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR