(6-Bromo-2-chloro-3-ethoxyphenyl)methanol 95%

Lieferant: Apollo Scientific

OR71010-1G OR71010-500MG
APOSOR71010-1GEA 278 EUR
APOSOR71010-1G APOSOR71010-500MG
(6-Bromo-2-chloro-3-ethoxyphenyl)methanol 95%
(6-Bromo-2-chloro-3-ethoxyphenyl)methanol
Formel: C₉H₁₀BrClO₂
Molecular Weight: 265,531 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD32707277
CAS-Nummer: 2386676-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR