1-(3-Aminoprop-1-yl)-1H-1,2,4-triazole hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Amino-3-(1H-1,2,4-triazol-1-yl)propane hydrochloride, 3-(1H-1,2,4-Triazol-1-yl)propylamine hydrochloride
OR7107-1G
OR7107-250MG
APOSOR7107-1GEA
219
EUR
APOSOR7107-1G
APOSOR7107-250MG
1-(3-Aminoprop-1-yl)-1H-1,2,4-triazole hydrochloride
1-(3-Aminoprop-1-yl)-1H-1,2,4-triazole hydrochloride
Formel:
C₅H₁₀N₄·ClH Molecular Weight: 162,621 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06796387 CAS-Nummer: 794522-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...