Iodobenzene dichloride 70%

Lieferant: Apollo Scientific

OR7132-1G OR7132-5G OR7132-250MG
APOSOR7132-1GEA 364 EUR
APOSOR7132-1G APOSOR7132-5G APOSOR7132-250MG
Iodobenzene dichloride 70%
Iodobenzene dichloride
Formel: C₆H₅Cl₂I
Molecular Weight: 274.914 g/mol
Schmelzpunkt: 110…112 °C (Zers.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00009608
CAS-Nummer: 932-72-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR