Methyl 4-bromo-2-nitrophenylacetate 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-2-(2-methoxy-2-oxoethyl)nitrobenzene, Methyl 2-(4-bromo-2-nitrophenyl)acetate, 4-Bromo-2-nitrophenylacetic acid methyl ester

OR7152-1G OR7152-5G
APOSOR7152-1GEA 48.2 EUR
APOSOR7152-1G APOSOR7152-5G
Methyl 4-bromo-2-nitrophenylacetate 97%
Methyl 4-bromo-2-nitrophenylacetate
Formel: C₉H₈BrNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09027267
CAS-Nummer: 100487-82-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR