4-(Cyclopropylmethoxy)benzeneboronic acid 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-(Cyclopropylmethoxy)phenyl)boronic acid, 4-(Cyclopropylmethoxy)phenylboronic acid
OR7194-1G
OR7194-10G
OR7194-25G
OR7194-100MG
OR7194-250MG
APOSOR7194-1GEA
159
EUR
APOSOR7194-1G
APOSOR7194-10G
APOSOR7194-25G
APOSOR7194-100MG
APOSOR7194-250MG
4-(Cyclopropylmethoxy)benzeneboronic acid 98%
4-(Cyclopropylmethoxy)benzeneboronic acid
Formel:
C₁₀H₁₃BO₃ Molecular Weight: 192.019 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04039017 CAS-Nummer: 411229-67-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...