1-Methyl-1H-indole-2-boronic acid, neopentyl glycol ester 96%
Lieferant: Apollo Scientific
Synonyme:
2-(5,5-Dimethyl-1,3,2-dioxaborinan-2-yl)-1-methyl-1H-indole
OR7237-1G
OR7237-5G
APOSOR7237-1GEA
238
EUR
APOSOR7237-1G
APOSOR7237-5G
1-Methyl-1H-indole-2-boronic acid, neopentyl glycol ester 96%
1-Methyl-1H-indole-2-boronic acid, neopentyl glycol ester
Formel:
C₁₄H₁₈BNO₂ Molecular Weight: 243,109 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06797198 CAS-Nummer: 905966-48-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...