2'-Amino-[1,1'-biphenyl]-4-carboxylic acid hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2'-Amino-4-carboxybiphenyl hydrochloride, 4-(2-Aminophenyl)benzoic acid hydrochloride

OR7316-1G OR7316-5G
APOSOR7316-1GEA 140 EUR
APOSOR7316-1G APOSOR7316-5G
2'-Amino-[1,1'-biphenyl]-4-carboxylic acid hydrochloride 95%
2'-Amino-[1,1'-biphenyl]-4-carboxylic acid hydrochloride
Formel: C₁₃H₁₁NO₂·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD07366471
CAS-Nummer: 1172328-87-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR