4'-Amino-[1,1'-biphenyl]-4-carboxylic acid hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

OR7320-1G OR7320-250MG
APOSOR7320-1GEA 190 EUR
APOSOR7320-1G APOSOR7320-250MG
4'-Amino-[1,1'-biphenyl]-4-carboxylic acid hydrochloride 95%
4'-Amino-[1,1'-biphenyl]-4-carboxylic acid hydrochloride
Formel: C₁₃H₁₁NO₂·ClH
Molecular Weight: 249.693 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08276229
CAS-Nummer: 41567-82-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR