2'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(2-Chlorophenyl)benzaldehyde, 2-Chloro-4'-formylbiphenyl

OR7346-1G OR7346-5G
APOSOR7346-1GEA 97.3 EUR
APOSOR7346-1G APOSOR7346-5G
2'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde 95%
2'-Chloro-[1,1'-biphenyl]-4-carboxaldehyde
Formel: C₁₃H₉ClO
Molecular Weight: 216.663 g/mol
Schmelzpunkt: 70…71 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631914
CAS-Nummer: 39802-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR