2-Hydrazino-5-methyl-3-nitropyridine 98%

Lieferant: Apollo Scientific

OR7353-1G OR7353-5G
APOSOR7353-1GEA 87.3 EUR
APOSOR7353-1G APOSOR7353-5G
2-Hydrazino-5-methyl-3-nitropyridine 98%
2-Hydrazino-5-methyl-3-nitropyridine
Formel: C₆H₈N₄O₂
Molecular Weight: 168,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09027277
CAS-Nummer: 21901-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR