2-(Fur-2-yl)aniline hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Aminophenyl)furan hydrochloride

OR7392-250MG OR7392-1G
APOSOR7392-250MGEA 147 EUR
APOSOR7392-250MG APOSOR7392-1G
2-(Fur-2-yl)aniline hydrochloride 98%
2-(Fur-2-yl)aniline hydrochloride
Formel: C₁₀H₉NO·ClH
Molecular Weight: 195,645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08445548
CAS-Nummer: 855948-13-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR