3-(Fur-2-yl)benzoic acid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Furan-2-yl)benzoic acid
OR7400-250MG
OR7400-1G
APOSOR7400-250MGEA
94.4
EUR
APOSOR7400-250MG
APOSOR7400-1G
3-(Fur-2-yl)benzoic acid 97%
3-(Fur-2-yl)benzoic acid
Formel:
C₁₁H₈O₃ Molecular Weight: 188,179 g/mol Schmelzpunkt: 157…158 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02690979 CAS-Nummer: 35461-99-5 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...