6-Bromo-1-methyl-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Bromo-1-methyl-1H-benzo[d]imidazole

OR7414-1G OR7414-5G
APOSOR7414-1GEA 60.9 EUR
APOSOR7414-1G APOSOR7414-5G
6-Bromo-1-methyl-1H-benzimidazole
6-Bromo-1-methyl-1H-benzimidazole
Formel: C₈H₇BrN₂
Molecular Weight: 211,059 g/mol
Schmelzpunkt: 201…203 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09027271
CAS-Nummer: 53484-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR