3-(Thien-2-yl)benzaldehyde
Lieferant: Apollo Scientific
Synonyme:
3-(Thiophen-2-yl)benzaldehyde
OR7589-1G
OR7589-5G
APOSOR7589-1GEA
183
EUR
APOSOR7589-1G
APOSOR7589-5G
3-(Thien-2-yl)benzaldehyde
3-(Thien-2-yl)benzaldehyde
Formel:
C₁₁H₈OS Molecular Weight: 188,246 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04038019 CAS-Nummer: 103668-99-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...