3-(Thien-3-yl)benzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3-Formylphenyl)thiophene, 3-(Thiophen-3-yl)benzaldehyde
OR7591-1G
OR7591-250MG
APOSOR7591-1GEA
181
EUR
APOSOR7591-1G
APOSOR7591-250MG
3-(Thien-3-yl)benzaldehyde
3-(Thien-3-yl)benzaldehyde
Formel:
C₁₁H₈OS Molecular Weight: 188,246 g/mol Schmelzpunkt: 45…46 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04039150 CAS-Nummer: 129746-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...