2-(Thien-2-yl)benzoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Carboxyphenyl)thiophene

OR7593-1G OR7593-250MG
APOSOR7593-1GEA 395 EUR
APOSOR7593-1G APOSOR7593-250MG
2-(Thien-2-yl)benzoic acid
2-(Thien-2-yl)benzoic acid
Formel: C₁₁H₈O₂S
Schmelzpunkt: 92…95 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04039152
CAS-Nummer: 6072-49-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR