2-Amino-3-(1,2-dihydro-2-oxoquinolin-4-yl)propanoic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-3-(1,2-dihydro-2-oxoquinolin-4-yl)propionic acid, 3-(1,2-Dihydro-2-oxoquinolin-4-yl)-DL-alanine

OR7606-25G OR7606-5G
APOSOR7606-25GEA 277 EUR
APOSOR7606-25G APOSOR7606-5G
2-Amino-3-(1,2-dihydro-2-oxoquinolin-4-yl)propanoic acid
2-Amino-3-(1,2-dihydro-2-oxoquinolin-4-yl)propanoic acid
Formel: C₁₂H₁₂N₂O₃
Molecular Weight: 232,235 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039159
CAS-Nummer: 5162-90-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR