5-Bromobenzo[b]thiophene-2-boronic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-1-benzothiophene-2-boronic acid, 2-Borono-5-bromobenzo[b]thiophene

OR7702-1G OR7702-5G
APOSOR7702-1GEA 143 EUR
APOSOR7702-1G APOSOR7702-5G
5-Bromobenzo[b]thiophene-2-boronic acid
5-Bromobenzo[b]thiophene-2-boronic acid
Formel: C₈H₆BBrO₂S
Molecular Weight: 256,912 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06801728
CAS-Nummer: 884010-25-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR