4-Bromo-3-[(dimethylamino)methyl]phenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-3-((dimethylamino)methyl)phenol, 1-(2-Bromo-5-hydroxyphenyl)-N,N-dimethylmethylamine, 2-Bromo-N,N-dimethyl-5-hydroxybenzylamine
OR7720-1G
OR7720-5G
APOSOR7720-1GEA
105
EUR
APOSOR7720-1G
APOSOR7720-5G
4-Bromo-3-[(dimethylamino)methyl]phenol
4-Bromo-3-[(dimethylamino)methyl]phenol
Formel:
C₉H₁₂BrNO Lagertemperatur: Kühlschrank |
MDL:
MFCD06657798 CAS-Nummer: 848848-16-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...