[(4-Bromophenyl)sulphonyl]acetonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromophenyl cyanomethyl sulphone
OR7778-5G
OR7778-25G
APOSOR7778-5GEA
128
EUR
APOSOR7778-5G
APOSOR7778-25G
[(4-Bromophenyl)sulphonyl]acetonitrile
[(4-Bromophenyl)sulphonyl]acetonitrile
Formel:
C₈H₆BrNO₂S Molecular Weight: 260,108 g/mol Schmelzpunkt: 190…192 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129455 CAS-Nummer: 126891-45-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...