3-[(4,6-Dimethoxypyrimidin-2-yl)oxy]pyridine-2-carboxylic acid monohydrate 95+%

Lieferant: Apollo Scientific

OR7847-1G OR7847-5G OR7847-10G
APOSOR7847-1GEA 185 EUR
APOSOR7847-1G APOSOR7847-5G APOSOR7847-10G
3-[(4,6-Dimethoxypyrimidin-2-yl)oxy]pyridine-2-carboxylic acid monohydrate 95+%
3-[(4,6-Dimethoxypyrimidin-2-yl)oxy]pyridine-2-carboxylic acid monohydrate
Formel: C₁₂H₁₁N₃O₅·H₂O
Molecular Weight: 295,248 g/mol
Schmelzpunkt: >130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08567951
CAS-Nummer: 1172989-15-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR