1H-Indene-3-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

OR7859-5G OR7859-10G
APOSOR7859-5GEA 219 EUR
APOSOR7859-5G APOSOR7859-10G
1H-Indene-3-carboxylic acid 95%
1H-Indene-3-carboxylic acid
Formel: C₁₀H₈O₂
Molecular Weight: 160,17 g/mol
Schmelzpunkt: 161…165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00086207
CAS-Nummer: 14209-41-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR