2-Acetyl-3-amino-5-(4-tert-butylphenyl)thiophene 97%

Lieferant: Apollo Scientific

OR8003-10G OR8003-1G OR8003-5G
APOSOR8003-10GEA 519 EUR
APOSOR8003-10G APOSOR8003-1G APOSOR8003-5G
2-Acetyl-3-amino-5-(4-tert-butylphenyl)thiophene 97%
2-Acetyl-3-amino-5-(4-tert-butylphenyl)thiophene
Formel: C₁₆H₁₉NOS
Molecular Weight: 273,393 g/mol
Schmelzpunkt: 179…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179917
CAS-Nummer: 306935-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR