2,1-Benzisoxazole-3-carboxylic acid 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzo[c]isoxazole-3-carboxylic acid, 3-Carboxy-2,1-benzisoxazole

OR8019-1G OR8019-5G OR8019-100MG
APOSOR8019-1GEA 246 EUR
APOSOR8019-1G APOSOR8019-5G APOSOR8019-100MG
2,1-Benzisoxazole-3-carboxylic acid 96%
2,1-Benzisoxazole-3-carboxylic acid
Formel: C₈H₅NO₃
Molecular Weight: 163.13 g/mol
Schmelzpunkt: 229…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02180526
CAS-Nummer: 642-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR