4-Chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine 97%

Lieferant: Apollo Scientific
Achtung

OR8057-10G OR8057-1G
APOSOR8057-10GEA 855 EUR
APOSOR8057-10G APOSOR8057-1G
4-Chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine 97%
4-Chloro-5-methyl-6-phenylthieno[2,3-d]pyrimidine
Formel: C₁₃H₉ClN₂S
Molecular Weight: 260,742 g/mol
Schmelzpunkt: 133…135 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02179828
CAS-Nummer: 306934-78-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR