2-(5-Chloro-1,2,4-thiadiazol-3-yl)thiophene-5-sulphonyl chloride 95%

Lieferant: Apollo Scientific
Gefahr

OR8065-1G OR8065-500MG
APOSOR8065-1GEA 458 EUR
APOSOR8065-1G APOSOR8065-500MG
2-(5-Chloro-1,2,4-thiadiazol-3-yl)thiophene-5-sulphonyl chloride 95%
2-(5-Chloro-1,2,4-thiadiazol-3-yl)thiophene-5-sulphonyl chloride
Formel: C₆H₂Cl₂N₂O₂S₃
Molecular Weight: 301.193 g/mol
Schmelzpunkt: 71…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02180888
CAS-Nummer: 306937-21-3
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR