Diethyl 3,4-dihydroxyfuran-2,5-dicarboxylate 97%
Lieferant: Apollo Scientific
Synonyme:
2,5-Bis(ethoxycarbonyl)-3,4-dihydroxyfuran
OR8077-1G
OR8077-250MG
APOSOR8077-1GEA
183
EUR
APOSOR8077-1G
APOSOR8077-250MG
Diethyl 3,4-dihydroxyfuran-2,5-dicarboxylate 97%
Diethyl 3,4-dihydroxyfuran-2,5-dicarboxylate
Formel:
C₁₀H₁₂O₇ Molecular Weight: 244,198 g/mol Schmelzpunkt: 178…180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00092386 CAS-Nummer: 6270-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...