Ethyl 1-(3-aminobenzyl)piperidine-4-carboxylate 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1-(3-Aminobenzyl)-4-(ethoxycarbonyl)piperidine, 3-{[4-Ethoxycarbonyl)piperidin-1-yl]methyl}aniline

OR8090-1G OR8090-250MG OR8090-5G
APOSOR8090-1GEA 102 EUR
APOSOR8090-1G APOSOR8090-250MG APOSOR8090-5G
Ethyl 1-(3-aminobenzyl)piperidine-4-carboxylate 97%
Ethyl 1-(3-aminobenzyl)piperidine-4-carboxylate
Formel: C₁₅H₂₂N₂O₂
Molecular Weight: 262,347 g/mol
Schmelzpunkt: 49…53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02180894
CAS-Nummer: 306937-22-4
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR