2-Methyl-5-(piperidin-1-ylsulphonyl)-3-furoic acid 95%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Methyl-5-(piperidin-1-ylsulphonyl)furan-3-carboxylic acid
OR8133-1G
OR8133-250MG
APOSOR8133-1GEA
140
EUR
APOSOR8133-1G
APOSOR8133-250MG
2-Methyl-5-(piperidin-1-ylsulphonyl)-3-furoic acid 95%
2-Methyl-5-(piperidin-1-ylsulphonyl)-3-furoic acid
Formel:
C₁₁H₁₅NO₅S Molecular Weight: 273,306 g/mol Schmelzpunkt: 188…189 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02180759 CAS-Nummer: 306936-38-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...