2-(4-Methyl-1,3-thiazol-2-yl)acetamide 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Amino-2-oxoethyl)-4-methyl-1,3-thiazole

OR8136-1G OR8136-250MG OR8136-5G
APOSOR8136-1GEA 313 EUR
APOSOR8136-1G APOSOR8136-250MG APOSOR8136-5G
2-(4-Methyl-1,3-thiazol-2-yl)acetamide 95%
2-(4-Methyl-1,3-thiazol-2-yl)acetamide
Formel: C₆H₈N₂OS
Molecular Weight: 156,206 g/mol
Schmelzpunkt: 128…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02180665
CAS-Nummer: 173739-73-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR