(Naphth-2-yl)acetyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-[(Chlorocarbonyl)methyl]naphthalene, 2-(Naphthalen-2-yl)acetyl chloride, 2-(Naphth-2-yl)ethanoyl chloride
OR8139-10G
OR8139-1G
OR8139-250MG
APOSOR8139-10GEA
377
EUR
APOSOR8139-10G
APOSOR8139-1G
APOSOR8139-250MG
(Naphth-2-yl)acetyl chloride
(Naphth-2-yl)acetyl chloride
Formel:
C₁₂H₉ClO Molecular Weight: 204,652 g/mol Schmelzpunkt: 60…61 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02180524 CAS-Nummer: 37859-25-9 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...