4-(5-Bromopent-1-yl)anisole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-(5-Bromopent-1-yl)-4-methoxybenzene, 1-Bromo-5-(4-methoxyphenyl)pentane
OR8175-25G
OR8175-5G
APOSOR8175-25GEA
716
EUR
APOSOR8175-25G
APOSOR8175-5G
4-(5-Bromopent-1-yl)anisole
4-(5-Bromopent-1-yl)anisole
Formel:
C₁₂H₁₇BrO Molecular Weight: 257.167 g/mol Siedepunkt: 123…125 °C (0.15 mm Hg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075176 CAS-Nummer: 14469-84-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...