7-(Benzyloxy)quinolin-4-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 7-(Benzyloxy)-4-hydroxyquinoline, 7-(Benzyloxy)-4-hydroxy-1-azanaphthalene

OR8291-1G OR8291-5G
APOSOR8291-1GEA 108 EUR
APOSOR8291-1G APOSOR8291-5G
7-(Benzyloxy)quinolin-4-ol
7-(Benzyloxy)quinolin-4-ol
Formel: C₁₆H₁₃NO₂
Molecular Weight: 251,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659963
CAS-Nummer: 749922-34-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR