4-Bromo-2-methyl-1,3-thiazole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-2-methylthiazole

OR8527-1G OR8527-250MG OR8527-5G
APOSOR8527-1GEA 87 EUR
APOSOR8527-1G APOSOR8527-250MG APOSOR8527-5G
4-Bromo-2-methyl-1,3-thiazole 95%
4-Bromo-2-methyl-1,3-thiazole
Formel: C₄H₄BrNS
Molecular Weight: 178.05 g/mol
Siedepunkt: 88…99 °C (16 mm Hg)
Lagertemperatur: Gefrierschrank
MDL: MFCD06409945
CAS-Nummer: 298694-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR