4'-(Piperidin-4-ylcarbonyl)acetanilide 97%
Lieferant: Apollo Scientific
Synonyme:
N-[4-(Piperidin-4-ylcarbonyl)phenyl]acetamide
OR8698-1G
OR8698-250MG
APOSOR8698-1GEA
219
EUR
APOSOR8698-1G
APOSOR8698-250MG
4'-(Piperidin-4-ylcarbonyl)acetanilide 97%
4'-(Piperidin-4-ylcarbonyl)acetanilide
Formel:
C₁₄H₁₈N₂O₂ Molecular Weight: 246.305 g/mol Schmelzpunkt: 167…170.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08690322 CAS-Nummer: 124035-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...