Methyl thiazolidine-2-carboxylate hydrochloride 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Thiazolidine-2-carboxylic acid methyl ester hydrochloride

OR8944-5G OR8944-100G OR8944-25G
APOSOR8944-5GEA 31.9 EUR
APOSOR8944-5G APOSOR8944-100G APOSOR8944-25G
Methyl thiazolidine-2-carboxylate hydrochloride 98%
Methyl thiazolidine-2-carboxylate hydrochloride
Formel: C₅H₉NO₂S·HCl
Molecular Weight: 183,656 g/mol
Schmelzpunkt: 158 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00068022
CAS-Nummer: 33305-08-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR